ENA018110095 Enamine

5-[(4-chlorophenoxy)methyl]-4-[(furan-2-yl)methyl]-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD04617313
  • Cas Number : 727696-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-07033

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07033
Availability details for
Quantity
  • Availability
  • Add to Cart