ENA018110096 Enamine

N,N-diethyl-6-hydrazinylpyridine-3-sulfonamide

  • MDL number MFCD04618154
  • Cas Number : 627837-60-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07034

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07034
Availability details for
Quantity
  • Availability
  • Add to Cart