ENA018110100 Enamine

N-(4-methoxy-2-nitrophenyl)-2-(3-oxo-1,2,3,4-tetrahydroquinoxalin-2-yl)acetamide

  • MDL number MFCD02926924
  • Cas Number : 476410-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07038

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07038
Availability details for
Quantity
  • Availability
  • Add to Cart