ENA018110101 Enamine

4-(2-chloroacetyl)-3,3-dimethyl-1,2,3,4-tetrahydroquinoxalin-2-one

  • MDL number MFCD04619485
  • Cas Number : 727717-71-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-171
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-07039

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07039
Availability details for
Quantity
  • Availability
  • Add to Cart