ENA018110104 Enamine

ethyl 3-(2-chloroacetamido)-5-(4-fluorophenyl)thiophene-2-carboxylate

  • MDL number MFCD04626492
  • Cas Number : 731011-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07042

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07042
Availability details for
Quantity
  • Availability
  • Add to Cart