ENA018110109 Enamine

8,8-dioxo-8lambda6-thia-1,9-diazatricyclo[8.5.0.0,2,7]pentadeca-2(7),3,5,9-tetraene-5-carboxylic acid

  • MDL number MFCD04624234
  • Cas Number : 730992-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07052

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07052
Availability details for
Quantity
  • Availability
  • Add to Cart