ENA018110112 Enamine

1-(4-bromophenyl)butane-1,3-dione

  • MDL number MFCD04619579
  • Cas Number : 4023-81-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92...94
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07055

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07055
Availability details for
Quantity
  • Availability
  • Add to Cart