ENA018110113 Enamine

1-(4-methylphenyl)butane-1,3-dione

  • MDL number MFCD00068483
  • Cas Number : 4023-79-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 19...21
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07056

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07056
Availability details for
Quantity
  • Availability
  • Add to Cart