ENA018110126 Enamine

4-(methylsulfanyl)-2-{[3-(morpholine-4-sulfonyl)phenyl]formamido}butanoic acid

  • MDL number MFCD04623045
  • Cas Number : 1008265-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07071

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07071
Availability details for
Quantity
  • Availability
  • Add to Cart