ENA018110133 Enamine

3-[2-(3,4-dimethoxyphenyl)ethyl]-2-mercapto-6-phenylthieno[2,3-d]pyrimidin-4(3H)-one

  • MDL number MFCD03945655
  • Cas Number : 590352-93-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07078

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07078
Availability details for
Quantity
  • Availability
  • Add to Cart