ENA018110156 Enamine

1-{4-hydroxy-3-[(pyrimidin-2-ylsulfanyl)methyl]phenyl}ethan-1-one

  • MDL number MFCD04624235
  • Cas Number : 730992-37-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07114

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07114
Availability details for
Quantity
  • Availability
  • Add to Cart