ENA018110167 Enamine

2-chloro-1-methyl-1H-indole-3,5-dicarbaldehyde

  • MDL number MFCD04624845
  • Cas Number : 66335-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164-165
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07126

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07126
Availability details for
Quantity
  • Availability
  • Add to Cart