ENA018110193 Enamine

1-[(4-nitrophenyl)sulfonyl]piperidine-2-carboxylic acid

  • MDL number MFCD00718652
  • Cas Number : 498531-42-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 161...163
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07155

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07155
Availability details for
Quantity
  • Availability
  • Add to Cart