ENA018110204 Enamine

1-(2,4,6-trimethylbenzenesulfonyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD03722940
  • Molecular Weight : 297.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07166

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07166
Availability details for
Quantity
  • Availability
  • Add to Cart