ENA018110205 Enamine

1-[(4-methoxyphenyl)sulfonyl]pyrrolidine-2-carboxylic acid

  • MDL number MFCD01892448
  • Cas Number : 202751-29-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07167

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07167
Availability details for
Quantity
  • Availability
  • Add to Cart