ENA018110213 Enamine

2,6-dimethylquinoline-3-carboxylic acid

  • MDL number MFCD06253921
  • Cas Number : 610261-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174...176
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07176

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07176
Availability details for
Quantity
  • Availability
  • Add to Cart