ENA018110231 Enamine

N-[2-amino-1-(4-methoxyphenyl)ethyl]-N,N-dimethylamine

  • MDL number MFCD01631936
  • Cas Number : 851169-57-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...162
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07196

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07196
Availability details for
Quantity
  • Availability
  • Add to Cart