ENA018110232 Enamine

2-(4-methoxyphenyl)-2-(morpholin-4-yl)ethan-1-amine

  • MDL number MFCD01631946
  • Cas Number : 31466-47-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07197

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07197
Availability details for
Quantity
  • Availability
  • Add to Cart