ENA018110244 Enamine

6-amino-5-(chloroacetyl)-1-propylpyrimidine-2,4(1H,3H)-dione

  • MDL number MFCD05263146
  • Cas Number : 743444-45-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07209

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07209
Availability details for
Quantity
  • Availability
  • Add to Cart