ENA018110256 Enamine

2-{[5-(2-fluorophenyl)-4-methyl-4H-1,2,4-triazol-3-yl]sulfanyl}acetic acid

  • MDL number MFCD04036356
  • Cas Number : 731003-94-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-07221

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07221
Availability details for
Quantity
  • Availability
  • Add to Cart