ENA018110257 Enamine

2-chloro-1-(2-methyl-1H-indol-3-yl)propan-1-one

  • MDL number MFCD04625526
  • Cas Number : 731003-87-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07223

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07223
Availability details for
Quantity
  • Availability
  • Add to Cart