ENA018110262 Enamine

{[(4-bromo-2-chlorophenyl)sulfonyl]amino}acetic acid

  • MDL number MFCD04625527
  • Cas Number : 731003-88-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07228

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07228
Availability details for
Quantity
  • Availability
  • Add to Cart