ENA018110267 Enamine

N-methyl-N-(4-methylbenzyl)amine

  • MDL number MFCD04115412
  • Cas Number : 699-04-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148...150
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07233

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-07233
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart