ENA018110269 Enamine

2-chloro-1-[1-(2,3-dihydro-1,4-benzodioxin-6-yl)-2,5-dimethyl-1H-pyrrol-3-yl]ethan-1-one

  • MDL number MFCD04625518
  • Cas Number : 731003-81-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100-101
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07235

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07235
Availability details for
Quantity
  • Availability
  • Add to Cart