ENA018110281 Enamine

2-(4-acetylphenoxy)-1-(pyrrolidin-1-yl)ethan-1-one

  • MDL number MFCD01673208
  • Cas Number : 42018-32-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96-97
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07248

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07248
Availability details for
Quantity
  • Availability
  • Add to Cart