ENA018110289 Enamine

5-(3-bromophenyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD04627325
  • Cas Number : 109060-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 239-240
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07256

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07256
Availability details for
Quantity
  • Availability
  • Add to Cart