ENA018110297 Enamine

3-(ethoxymethyl)-1-benzofuran-2-carboxylic acid

  • MDL number MFCD04627328
  • Cas Number : 731802-26-9
  • Molecular Weight : 220.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95-96
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07265

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07265
Availability details for
Quantity
  • Availability
  • Add to Cart