ENA018110298 Enamine

4-[(1,2,3,4-thiatriazol-5-yl)amino]phenol

  • MDL number MFCD00003131
  • Cas Number : 23567-67-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07267

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07267
s_phrases_eu 26-37/39
Availability details for
Quantity
  • Availability
  • Add to Cart