ENA018110301 Enamine

2-cyano-3-(1-cyclohexyl-2,5-dimethyl-1H-pyrrol-3-yl)prop-2-enoic acid

  • Cas Number : 732291-33-7
  • Molecular Weight : 272.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-07270

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07270
Availability details for
Quantity
  • Availability
  • Add to Cart