ENA018110304 Enamine

ethyl 2-(benzylamino)-5-methyl-1,3-thiazole-4-carboxylate

  • MDL number MFCD04627318
  • Cas Number : 731802-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-07273

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07273
Availability details for
Quantity
  • Availability
  • Add to Cart