ENA018110309 Enamine

methyl({[4-(propan-2-yl)phenyl]methyl})amine

  • MDL number MFCD04627902
  • Cas Number : 73441-51-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07278

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07278
Availability details for
Quantity
  • Availability
  • Add to Cart