ENA018110317 Enamine

ethyl 2-[(chloroacetyl)amino]-4-methyl-5-phenylthiophene-3-carboxylate

  • MDL number MFCD04628439
  • Cas Number : 731820-85-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 106...108
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07287

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07287
Availability details for
Quantity
  • Availability
  • Add to Cart