ENA018110318 Enamine

3-[2-(3,4-dimethoxyphenyl)ethyl]-6-ethyl-2-sulfanyl-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD03944343
  • Cas Number : 588682-61-5
  • Molecular Weight : 376.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07288

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07288
Availability details for
Quantity
  • Availability
  • Add to Cart