ENA018110320 Enamine

4-amino-5-(3-chlorophenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD01940466
  • Cas Number : 117320-66-8
  • Molecular Weight : 226.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-07290

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07290
Availability details for
Quantity
  • Availability
  • Add to Cart