ENA018110324 Enamine

(3,4-dimethylphenyl)thiourea

  • MDL number MFCD00060452
  • Cas Number : 16738-18-4
  • Molecular Weight : 180.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-07294

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07294
Availability details for
Quantity
  • Availability
  • Add to Cart