ENA018110327 Enamine

methyl 5-(2-chloropropanoyl)-2,4-dimethyl-1H-pyrrole-3-carboxylate

  • MDL number MFCD04627903
  • Cas Number : 731815-67-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07297

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07297
Availability details for
Quantity
  • Availability
  • Add to Cart