ENA018110355 Enamine

2-chloro-N-(3,5-dimethyl-1H-pyrazol-4-yl)acetamide

  • MDL number MFCD03274584
  • Cas Number : 774175-89-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145-146
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07328

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07328
Availability details for
Quantity
  • Availability
  • Add to Cart