ENA018110365 Enamine

4-chloro-3-({[(1-methyl-1H-imidazol-2-yl)thio]acetyl}amino)benzoic acid

  • MDL number MFCD04631454
  • Cas Number : 731793-31-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 223...225
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07341

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07341
Availability details for
Quantity
  • Availability
  • Add to Cart