ENA018110376 Enamine

1-[4-fluoro-2-(pyrrolidin-1-yl)phenyl]ethan-1-one

  • MDL number MFCD05263169
  • Cas Number : 743444-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07352

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07352
Availability details for
Quantity
  • Availability
  • Add to Cart