ENA018110378 Enamine

1-(1-benzyl-2,5-dimethyl-1H-pyrrol-3-yl)-2-chloroethanone

  • MDL number MFCD04629634
  • Cas Number : 610274-26-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65...67
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07356

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07356
Availability details for
Quantity
  • Availability
  • Add to Cart