ENA018110384 Enamine

4-nitro-N1-phenylbenzene-1,2-diamine

  • MDL number MFCD00234791
  • Cas Number : 55315-12-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-07363

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07363
Availability details for
Quantity
  • Availability
  • Add to Cart