ENA018110390 Enamine

5-[(2-methoxyethyl)amino]-1,3,4-thiadiazole-2-thiol

  • MDL number MFCD04629629
  • Cas Number : 731776-65-1
  • Molecular Weight : 191.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-121
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-07370

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07370
Availability details for
Quantity
  • Availability
  • Add to Cart