ENA018110393 Enamine

2-(2-bromo-4,6-difluorobenzenesulfonamido)acetic acid

  • MDL number MFCD04629612
  • Cas Number : 731776-56-0
  • Molecular Weight : 330.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-07373

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07373
Availability details for
Quantity
  • Availability
  • Add to Cart