ENA018110401 Enamine

3-methyl-1-[(naphthalen-1-yl)methyl]-5-(pyrrolidin-1-yl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD05263205
  • Cas Number : 926189-81-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07382

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07382
Availability details for
Quantity
  • Availability
  • Add to Cart