ENA018110405 Enamine

ethyl 1-ethyl-6-methyl-2-sulfanylidene-4-(thiophen-2-yl)-1,2,3,4-tetrahydropyrimidine-5-carboxylate

  • MDL number MFCD05263207
  • Cas Number : 743444-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07386

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07386
Availability details for
Quantity
  • Availability
  • Add to Cart