ENA018110431 Enamine

3-chloro-5-(pyridin-3-yl)-8-thia-4,6-diazatricyclo[7.4.0.0,2,7]trideca-1(9),2,4,6-tetraene

  • MDL number MFCD00705137
  • Cas Number : 128277-24-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100-101
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-07412

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07412
Availability details for
Quantity
  • Availability
  • Add to Cart