ENA018110436 Enamine

3-{4-chloro-5-phenylthieno[2,3-d]pyrimidin-2-yl}pyridine

  • MDL number MFCD05263811
  • Cas Number : 743453-38-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07417

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07417
Availability details for
Quantity
  • Availability
  • Add to Cart