ENA018110442 Enamine

methyl 4-chloro-2,5-dimethylthieno[2,3-d]pyrimidine-6-carboxylate

  • MDL number MFCD06660679
  • Cas Number : 872319-75-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07423

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07423
Availability details for
Quantity
  • Availability
  • Add to Cart