ENA018110456 Enamine

N-(4-ethoxy-3-methoxybenzyl)-N-methylamine

  • MDL number MFCD04622320
  • Cas Number : 709649-58-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 19...21
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07480

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07480
Availability details for
Quantity
  • Availability
  • Add to Cart