ENA018110457 Enamine

N-methyl-N-(2,3,4-trimethoxybenzyl)amine

  • MDL number MFCD04622289
  • Cas Number : 709649-63-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 127...130
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07481

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07481
Availability details for
Quantity
  • Availability
  • Add to Cart